Reducing Union, Parker, Restek
Supplier: Restek
21825 21925 22869 21923 22523 22524 22525
RK21825PK 225.83 CAD
RK21825 10854-488 10854-760 10854-762 10854-960 10854-962 10855-014 10855-296 10855-402 10855-486 10855-488 10855-554
Reducing Union, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Available with connection sizes of 1/16", 1/8", or 1/4" in diameter.