Port Connector, Parker, Restek
Supplier: Avantor
21550 21548 21549
10854-490EA 132.89 CAD
10854-490 10855-184 10855-246 10855-248 10855-428 10855-590 10855-604 10855-606 10855-650 10855-652
Port Connector, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Available with connection sizes of 1/8" or 1/4" in diameter.