Plug, Parker, Restek
Supplier: Restek
21816 21916 21917 21817 21540
RK21816PK 221.3 CAD
RK21816 10854-930 10854-946 10854-948 10855-006 10855-008 10855-608 10855-610 10855-816 10855-818
Plug, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Available in sizes of 1/16", 1/8", or 1/4" in diameter.
Order Now
SPECIFICATIONS
Environmentally Preferable