Nut, Parker, Restek
Supplier: Restek
21900 21901 21902 21801 21802
RK21900PK 115.07 CAD
RK21900 10854-942 10854-944 10855-644 10855-646 10855-716
Nut, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
No coating required, but nuts can be treated upon request (custom parts).
Ordering information: Note: Nuts and ferrules are not treated unless requested (custom parts). Nuts and ferrules normally are not in contact with sample pathway, and thus do not require coating.