Male Quick Coupling, Parker, Restek
Supplier: Restek
21957
10854-492EA 252.71 CAD
10854-492 10854-494 10855-532 10855-534
Male Quick Coupling, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Can be treated with Siltek®/Sulfinert or Silcosteel®-CR
Compatible with 1/4" or 1/8" openings depending on model.