Front Ferrule, Parker, Restek
Supplier: Restek
21904 21905 21903 21803 21804 21805
10854-886EA 162.9 CAD
10854-886 10854-888 10855-366 10855-424 10855-478 10855-480
Front Ferrule, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
No coating required, but ferrules can be treated upon request (custom parts).
Ordering information: Note: Nuts and ferrules are not treated unless requested (custom parts). Nuts and ferrules normally are not in contact with sample pathway, and thus do not require coating.