Elbow, Parker, Restek
Supplier: Restek
22530 22531
10855-780EA 241.4 CAD
10855-780 10855-782 10855-800 10855-802
Elbow, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Elbow fittings are hot forged and machined.