Straight Inlet Liner for Bruker/Varian GCs equipped with 1078/1079 inlets, Restek
Supplier: Restek
21715
75779-292PK 222.79 CAD
75779-292 75779-500 75779-510
Straight Inlet Liner for Bruker/Varian GCs equipped with 1078/1079 inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.