Straight Inlet Liner with CarboFrit® for Thermo TRACE GCs Equipped with PTV Inlets, Restek
Supplier: Restek
21117-216 21117-209
75778-434PK 573.14 CAD
75778-434 75778-950
Straight Inlet Liner with CarboFrit® for Thermo TRACE GCs Equipped with PTV Inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.