Baffled Inlet Liner for Thermo TRACE GCs equipped with PTV inlets, Restek
Supplier: Restek
22075 22075-214
75778-728PK 362.25 CAD
75778-728 75779-456
Baffled Inlet Liner for Thermo TRACE GCs equipped with PTV inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.