Straight Inlet Liners for Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets, Restek
Supplier: Restek
20937 20940 20917 22267 20940-214 20917-214
RK20937PK 273.6 CAD
RK20937 RK20940 RK20917 75779-172 75779-452 75779-454 75779-508
Straight Inlet Liners for Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Order Now
SPECIFICATIONS
Environmentally Preferable