Straight Inlet Liner with Wool for Shimadzu 17A GCs equipped with PTV inlets, Restek
Supplier: Avantor
RK21706PK 352.24 CAD
RK21706
Straight Inlet Liner with Wool for Shimadzu 17A GCs equipped with PTV inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.