Straight Inlet Liner with Wool for Shimadzu 17A, 2010, and 2014 GCs Equipped with Split/Splitless Inlets, Restek
Supplier: Restek
20956-213 20956-211 20956
RK20956-213.5PK 499.61 CAD
RK20956-213.5 75779-466 75779-518
Straight Inlet Liner with Wool for Shimadzu 17A, 2010, and 2014 GCs Equipped with Split/Splitless Inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.