Straight Inlet Liner for Agilent GCs equipped with Split/Splitless Inlets, Restek
Supplier: Restek
20713 20773 20973 20913 20915 20713-210 20782-210 20973-214 21700-205 20782-205 22976 20713-214 20773-210 20773-214 20973-210 20782-214
RK20713PK 275.23 CAD
RK20713 RK20773 RK20973 RK20913 RK20915 10803-470 10803-474 10803-484 75778-552 75778-638 75778-818 75778-956 75778-970 75779-382 75779-462 75779-492
Straight Inlet Liner for Agilent GCs equipped with Split/Splitless Inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.