Drilled Cyclo Uniliner® Inlet Liner with Hole near Top and Top Taper for Agilent GCs Equipped with Split/Splitless Inlets, Restek
Supplier: Restek
22980-214 22980
75778-122PK 853.79 CAD
75778-122 75778-560
Drilled Cyclo Uniliner® Inlet Liner with Hole near Top and Top Taper for Agilent GCs Equipped with Split/Splitless Inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.