Cup Inlet Liner with Wool for Agilent GCs Equipped with Split/Splitless Inlets, Restek
Supplier: Restek
20710-213.5 20710-211.5
75778-276PK 710.2 CAD
75778-276 75778-126
Cup Inlet Liner with Wool for Agilent GCs Equipped with Split/Splitless Inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.