Uniliner® Inlet Liner for Agilent GCs Equipped with Split/Splitless Inlets, Restek
Supplier: Restek
21053 21053-214
10803-236EA 659.87 CAD
10803-236 75779-042
Uniliner® Inlet Liner for Agilent GCs Equipped with Split/Splitless Inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.