Straight/SPME Inlet Liners for Agilent GCs equipped with Split/Splitless Inlets, Restek
Supplier: Restek
21111 21111-214 21111-210
10803-240EA 389.89 CAD
10803-240 75779-044 75779-334
Straight/SPME Inlet Liners for Agilent GCs equipped with Split/Splitless Inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.