The Claw and The Claw Holder Kit, Restek
Supplier: Restek
26262 26261
10852-858KT 125.94 CAD
10852-858 10852-904
The Claw and The Claw Holder Kit, Restek
Tool Sets
The Claw safely and cleanly removes liners, O-rings, or other small objects from the injection port, saving fingers from being burned while removing hot injection port liners.
After removal, you can then place the hot objects in a clean 4 mL vial situated in the Claw holder until ready for reuse.