Certified Autosampler Vial Kits, Certified 2.0 ml Screw-Thread Vials, Restek
Supplier: Restek
22788 22789 22790 22791 22792 22793
10058-790PK 312.85 CAD
10058-790 10058-792 10854-146 10854-148 10854-150 10854-152
Certified Autosampler Vial Kits, Certified 2.0 ml Screw-Thread Vials, Restek
Vials
Certified 2.0 ml screw-thread vial kits are available for optimal use with autosamplers.
  • Certificates enclosed for each kit
  • Clear or amber borosilicate glass fabrication
  • Closures with pre-inserted PTFE/silicone septa
Each kit is sealed for ensured integrity. Certificate parameters include vial dimensions and chemical test results.