ETP Electron Multipliers for Mass Spectrometry, Restek
Supplier: Avantor
23074 23075
10058-852EA 2788.98 CAD
10058-852 10058-854 10803-308
ETP Electron Multipliers for Mass Spectrometry, Restek
Chromatography Detectors
The electron multipliers manufactured by ETP use a proprietary dynode material that has a number of properties that make it very suitable for use in an electron multiplier.
  • Increased surface area for enhanced sensitivity and extended operational life
  • Optimized ion and electronic optics for maximum performance
It has very high secondary electron emission, which allows exceptional gain to be achieved from each dynode. This material is also very stable in air. In fact, an ETP multiplier can be stored for years before being used. As a direct result of theMore Product Information