Dual Vespel® Ring Cross-Disk Inlet Seals for Agilent GCs, Restek
Supplier: Restek
22086 22083 22084 22085
10803-192PK 848.19 CAD
10803-192 10855-930 10855-932 10855-934
Dual Vespel® Ring Cross-Disk Inlet Seals for Agilent GCs, Restek
Seals Mechanical Seals
Cross-disk inlet seals are treated to provide enhanced inertness for use in a variety of applications.
  • Washerless, leak-tight seal for Agilent GCs ensures better tightness and easier handling
  • Ring in top surface reduces torque required to seal
  • Ring in bottom surface eliminates need for a washer
Ideal for high-flow split applications, these inlet seals are gold-plated or Siltek®-treated.