Capillary Installation Gauge for Shimadzu 17A, 2010, and 2014 GCs, Restek
Supplier: Restek
22333
RK22333EA 159.6 CAD
RK22333
Capillary Installation Gauge for Shimadzu 17A, 2010, and 2014 GCs, Restek
Puller and Inserter Tools
Use a capillary installation gauge for Shimadzu 17A, 2010, and 2014 GCs to seat ferrules onto a column for consistent installations.
  • For use with graphite ferrules only
Ensure reliable, consistent ferrule installation with these capillary installation gauges.
Ordering information: For use with graphite ferrules only.