Liner Cap Removing Tool for Thermo Scientific GCs, Restek
Supplier: Restek
24937
10852-834EA 121.53 CAD
10852-834
Liner Cap Removing Tool for Thermo Scientific GCs, Restek
Puller and Inserter Tools
For use with Thermo Scientific Focus, TRACE, and TRACE Ultra GCs.
  • Easily loosens the liner cap from the injector
  • Unique, ergonomic handle that is easy to grip