Gap Inspection Gauge, Restek
Supplier: Restek
22624
10855-194EA 50.92 CAD
10855-194
Gap Inspection Gauge, Restek
Tool Sets
Gap inspection gauges are for Swagelok fittings in new installations only.
  • Confirm that fittings are sufficiently tightened
  • For use with 1/4", 3/8", 1/2" Swagelok fittings