EZ Twist Top® Split/Splitless Injection Port for Agilent GCs with Pencil Traps, Restek
Supplier: Restek
22721 22722
10856-062EA 1614.75 CAD
10856-062 10856-064
EZ Twist Top® Split/Splitless Injection Port for Agilent GCs with Pencil Traps, Restek
Tool Sets
With these tools, gas lines are attached to the shell weldment bottom to help eliminate broken gas lines.
  • Change inlet liners faster and easier
  • Gas lines don't interfere with routine maintenance
  • Weldment stays secure in the tool for reattachment
Weldment removal tool allows for quick removal of the hot weldment without fingers ever touching hot surfaces. For use with Agilent 6850/6890 GCs with pencil traps.