EZ Twist Top® Split/Splitless Injection Port for Agilent GCs, Restek
Supplier: Restek
22725 22726
10803-298EA 1487.37 CAD
10803-298 10856-174
EZ Twist Top® Split/Splitless Injection Port for Agilent GCs, Restek
Chromatography Supplies Chromatography Spare Parts
Using Restek’s unique EZ Twist Top® Injection Port and Restek Cool Tools (Weldment Removal Tool and Septum Nut Removal Tool), you can reduce maintenance time and frustration, and eliminate tangled gas lines and damage that leads to gas leaks, while avoiding direct contact with hot metal and glass surfaces.
Peak shape degradation, poor reproducibility, sample decomposition, and ghost peaks all are associated with using a dirty liner. Frequent septum replacement prevents fragmentation and leaks. The gas lines are attached to the EZ Twist Top® Shell Weldment (bottom) instead of the weldment (More Product Information