Click-On In-Line Super Clean® Replacement Gas Traps, Restek
Supplier: Restek
22464 22465
10854-566EA 462.79 CAD
10854-566 10855-630
Click-On In-Line Super Clean® Replacement Gas Traps, Restek
Filters Gas Filters
Super-clean click-on traps measure to be 21.6×3.2 cm (8.5×1.25").
  • Triple trap - removes oxygen, moisture, and hydrocarbons
Ordering information: Replacement trap only - fittings sold separately.