Capillary Installation Gauge for Thermo Scientific TRACE & Focus SSL, M4 Ferrules, Restek
Supplier: Restek
22330
10852-830EA 207.06 CAD
10852-830
Capillary Installation Gauge for Thermo Scientific TRACE & Focus SSL, M4 Ferrules, Restek
Puller and Inserter Tools
For use with Thermo Scientific TRACE & Focus SSL.
  • Seats ferrule onto column for consistent installations
  • Prevents crushed column ends
  • High-quality stainless steel construction
Ordering information: For use with graphite ferrules only.