ResTape PTFE Tape, Restek
Supplier: Restek
22486 22487
10058-782RL 26.14 CAD
10058-782 10852-824
ResTape PTFE Tape, Restek
Tapes
ResTape PTFE tape is for use with threaded connections in a wide range of plumbing materials.
  • Compatible with gaseous or liquid oxygen, and with many other gases and liquids
  • Anti-galling
  • Compatible with many other metals and polymers
Tapes have a maximum effective temperature of 260 °C.
Ordering information: Compatible with gaseous or liquid oxygen, and with many other gases and liquids. Compatible with a broad range of gases and liquids. Anti-galling. Also compatible with many other metals and polymers.