Straight Seamless 316L Grade Stainless Steel Tubing, Treated, Restek
Supplier: Restek
22901 22902 22903 22899 22900
10854-660EA 1172.59 CAD
10854-660 10854-662 10854-664 10854-858 10854-860 10854-902
Straight Seamless 316L Grade Stainless Steel Tubing, Treated, Restek
Tubing Transfer Tubing
Choose from 6-foot straight lengths of stainless steel tubing with outside diameters of 1/8" to 3/8".
  • Seamless stainless steel construction
  • Tubing can be cut to your exact requirements
  • Multiple lengths and different inside and outside diameters available
Lengths of stainless steel tubing are treated for optimal inertness and to extend lifetime.