Micropacked Inlet Conversion Kits, Restek
Supplier: Restek
22424 22426 22427 22428
10856-006KT 305.94 CAD
10856-006 10856-008 10856-010 10856-012 10856-014
Micropacked Inlet Conversion Kits, Restek
Fittings and Connectors
Use micropacked inlet conversion kits to convert capillary GC split/splitless inlets for use with 1/16" OD micropacked columns.
  • Sample pathways deactivated for ultimate inertness
For use with Agilent 5890, 6890, and 7890 GCs.
Accessories information: Replacement parts include dual Vespel ring inlet seals, and large bores (2).
Ordering information: For use with packed column FIDs only.