Deactivated Glass Wool, Restek
Supplier: Restek
24324
RK24324EA 117.99 CAD
RK24324
Deactivated Glass Wool, Restek
Glass and Cotton Wool
Use deactivated glass wool to vaporize a sample in a liner prior to introduction into a capillary column.
  • More inert than traditional glass wool