Septa Holder Kits for Thermo Scientific TRACE, 8000, 8000 TOP and Focus SSL, Restek
Supplier: Restek
21299 24972
10855-902EA 171.74 CAD
10855-902 10856-150
Septa Holder Kits for Thermo Scientific TRACE, 8000, 8000 TOP and Focus SSL, Restek
Chromatography Supplies Chromatography Spare Parts
High-quality stainless steel construction.
Silcosteel®-AC-treated version helps with septum removal.