Oven Temperature Sensor Assembly for Agilent GCs, Restek
Supplier: Restek
23039 23040
10500-580EA 231.18 CAD
10500-580 10856-092
Oven Temperature Sensor Assembly for Agilent GCs, Restek
Temperature Probes
The oven sensors works in conjunction with the oven control board to regulate oven temperature.