Replacement Needles for Agilent 7673, 7683, 7693A, and 6850 Autosampler Syringes, Hamilton, Restek
Supplier: Restek
20164 20165 20166
10059-064PK 216.83 CAD
10059-064 10059-066 10059-068
Replacement Needles for Agilent 7673, 7683, 7693A, and 6850 Autosampler Syringes, Hamilton, Restek
Syringe Needles
For use with Agilent 7673, 7683, 7693A, and 6850 autosamplers.
  • Replacement needle volume of 5 μL–10 μL
Order Now
SPECIFICATIONS
Environmentally Preferable