Vu2 Union® Connectors, Restek
Supplier: Restek
21105 21107 21106
RK21105EA 426.78 CAD
RK21105 10803-238 10852-978 10852-994
Vu2 Union® Connectors, Restek
Fittings and Connectors
Restek’s Vu2 Union® connector combines the simplicity of a Press-Tight® union with the strength of a metal union.
  • Open design allows visual confirmation of seal
  • Connector can be hung from column cage to minimize stress on the connections
  • Simple, reliable, easy-to-use solution for connection needs
A Press-Tight® union in the Vu2 Union® connector joins the fused silica tubing ends together; the ferrule and knurled nut at each end of the connector hold the tubing in place via a secondary seal between the ferrule and the Press-Tight® union. Each knurled nut applies independent pressure to each ferrule to make leak-tight seals with the column ends. These ultra-strong connections will not unexpectedly disconnect under temperature changes, vibrations, or other stresses normally encountered in GC analyses.
Ordering information: Kits include: Vu2 Union® body, two knurled nuts, two Press-Tight® unions, and four ferrules.
Note: This product is not recommended for GC column-to-MS connections.