Capillary Installation Gauge for Agilent 5973/5975 MS, Restek
Supplier: Restek
21894
10058-700EA 562.63 CAD
10058-700
Capillary Installation Gauge for Agilent 5973/5975 MS, Restek
Puller and Inserter Tools
Capillary installation gauges seat ferrules onto columns for consistent installations.
  • Made from high-quality stainless steel
For use with Agilent 5973/5975 MS.