Gas Pressure Gauge Kit, Restek
Supplier: Restek
21657
10855-642EA 187.37 CAD
10855-642
Gas Pressure Gauge Kit, Restek
Manometers
Use an in-line gas pressure gauge to indicate when a thermal gas purifier getter tube should be replaced.
Delivery information: Includes 1/8" tee and 0 to 100 psi (0 to 689 kPa) gauge.