PTFE Tip, Gas-Tight Syringes, SGE, Restek
Supplier: Restek
24742 24737
RK24742EA 138.73 CAD
RK24742 10059-254
PTFE Tip, Gas-Tight Syringes, SGE, Restek
Syringes Syringes for Chromatography
Interchangeable barrels, plungers, and tips extend performance and increase cost-effectiveness.
  • Suitable for gases or liquids
  • High accuracy of dispensed volumes
Ordering information: Needles sold separately.