Micro-Volume, Positive Displacement Syringes, SGE, Restek
Supplier: Restek
24771 24772 24775 24776 24778 24779
10059-266EA 248.62 CAD
10059-266 10059-268 10059-270 10059-272 10059-274 10059-276
Micro-Volume, Positive Displacement Syringes, SGE, Restek
Syringes Syringes for Chromatography
Sample is contained in the needle with full plunger displacement, ensuring no dead volume.
  • High precision and accuracy down to 0.1 µL
  • Replaceable needle/plunger kits and barrels (available on request)
Seal between the plunger and the needle can be tightened for high-pressure injection.