Replacement Plunger Assembly, PTFE-Tipped, for Agilent 7673, 7683, 7693A, and 6850 Autosampler Syringes, SGE, Restek
Supplier: Restek
21218 21284
10058-644PK 142.29 CAD
10058-644 76121-476
Replacement Plunger Assembly, PTFE-Tipped, for Agilent 7673, 7683, 7693A, and 6850 Autosampler Syringes, SGE, Restek
Syringe Barrels, Plungers and Pistons
For use with Agilent 7673, 7683, 7693A, and 6850 autosamplers.
  • PTFE-Tipped
  • Compatible with syringes with a volume of 10μL