Gas-Tight PTFE-Tipped Syringes for Agilent 7673, 7683, 7693A, and 6850 Autosamplers, Hamilton, Restek
Supplier: Restek
21224 21228 21229 24894
10058-648EA 212.82 CAD
10058-648 10058-650 10058-652 10059-318
Gas-Tight PTFE-Tipped Syringes for Agilent 7673, 7683, 7693A, and 6850 Autosamplers, Hamilton, Restek
Syringes Syringes for Chromatography
For use with Agilent 7673, 7683, 7693A, and 6850 autosamplers.
  • Gas-tight
  • PTFE-tipped