Super Clean® Gas Filter Kits and Replacements, Restek
Supplier: Restek
22019 22021 22031
RK22019EA 1597.23 CAD
RK22019 10142-090 10855-410
Super Clean® Gas Filter Kits and Replacements, Restek
Filters Gas Filters
Ensure high-purity output in these gas filters with 99.9999% pure gas (at a max flow of 2 L/min) with the use of these filter kits.
  • Designed for easy, hassle-free cartridge changes
  • Glass inside to prevent diffusion; polycarbonate housing outside for safety
  • All traps measure 27×4.4 cm (10⁵/₈×1³/₄")
  • Each base plate unit measures 10.2×10.2×4.8 cm (4×4×1⁷/₈")