Rethreading Tool, Restek
Supplier: Restek
23016 23017
10852-846EA 128.55 CAD
10852-846 10852-856 10852-864 10852-866
Rethreading Tool, Restek
Tool Sets
Rethreading tools repair worn or damaged threads that come about as a result of consistent use and temperature changes.
  • Multiple uses (fittings, injectors, detectors, etc.)
  • Built-in guide to prevent cross-threading
Repeated installation, removal, and temperature changes cause wear and damage to threads on GC parts. This can cause a poor seal, and oxygen can enter the system, compromising analytical results and possibly destroying expensive analytical columns.