Inlet Liner Seals for Bruker/Varian 1177 Injectors, Restek
Supplier: Restek
20297 20296 20299 20298
RK20297PK 360.35 CAD
RK20297 RK20296 RK20299 RK20298
Inlet Liner Seals for Bruker/Varian 1177 Injectors, Restek
Seals Mechanical Seals
Meets or exceeds original manufacturer's performance.
  • O-rings feature internal diameters of 6.35 mm or 6.5 mm depending on model
  • Max operating temperature of 450°C
  • Graphite
Inlet liner seals are fabricated from durable graphite.