Split/Splitless Injection Port for Agilent 6850/6890 GCs, Direct Replacement or Siltek® Treated, Restek
Supplier: Restek
22670 22671 22672 22674 20265 22673 20267 22686
RK22670EA 1270.08 CAD
RK22670 RK22671 RK22672 RK22674 RK20265 RK22673 10057-898 10057-914
Split/Splitless Injection Port for Agilent 6850/6890 GCs, Direct Replacement or Siltek® Treated, Restek
Chromatography Supplies Chromatography Spare Parts
Siltek®-treated split/splitless injector is a direct replacement for Agilent 5890 and 6850/6890 GCs, but Siltek® treatment passivates the metal surface to ensure an inert pathway for the sample, delivering increased performance.
  • Injector is manufactured from high-quality stainless steel
  • Meets or exceeds Agilent original equipment performance