Capillary Column Caps, Restek
Supplier: Restek
21044 22858
10852-844EA 226.64 CAD
10852-844 10852-878
Capillary Column Caps, Restek
Fittings and Connectors
Silicone or glass capillary column caps can be attached to the column in seconds to form an airtight seal.
  • Increase column lifetime—prevent moisture and air from entering the column during storage
  • Two styles to choose from: glass or silicone material
  • Glass caps are color-coded for identifying detector and injector ends
  • Not recommended for reuse