Coiled 316L Grade Stainless Steel Tubing, Treated, Restek
Supplier: Restek
29067 29097 29100 29103 29104 29114 29085 29087 29088 29108 29111 29112 29105 29092 29095 29096 29068 29089 29113 29107 29081 29073 29076 29078 29079 29080 29082 29083 29070 29071 29072 29075 29074 29098 29106 29090
10854-422RL 366.72 CAD
10854-422 10854-430 10854-432 10854-434 10854-436 10854-438 10854-440 10854-442 10854-544 10854-546 10854-548 10854-550 10854-552 10854-554 10854-556 10854-558 10854-560 10854-562 10854-580 10854-600 10854-602 10854-604 10854-606 10854-608 10854-610 10854-834 10855-038 10855-040 10855-042 10855-072 10855-074 10855-076 10855-078 10855-080 10855-082 10855-084 10855-088 10855-218 10855-250 10855-254 10855-256 10855-258 10855-260 10855-262 10855-266 10855-382 10855-384 10855-460
Coiled 316L Grade Stainless Steel Tubing, Treated, Restek
Tubing Transfer Tubing
Coiled tubing is available with inertness treatment for use in a variety of applications.
  • Stainless steel fabrication
  • Multiple lengths and different inside and outside diameters available
Seamless stainless steel tubing is differentiated by length and inside and outside diameter.
Ordering information: Maximum continuous lengths are: 1500' (1/8" Ext.Ø), 750 ft (1/4" Ext.Ø), and 250 ft (3/8" Ext.Ø). The availability of long lengths is subject to inventory constraints. Lead times may vary depending on the continuous length needed.