Autosampler Syringes for Thermo Scientific GCs, SGE, Restek
Supplier: Restek
24924 24934
10059-326EA 160.36 CAD
10059-326 10059-328
Autosampler Syringes for Thermo Scientific GCs, SGE, Restek
Syringes Syringes for Chromatography
Hamilton and SGE syringes are designed and tested to meet critical autosampler performance needs.
  • Needle point styles are designed to withstand multiple rapid injections through a septum
For use with Thermo Scientific GCs.